Product Details
- SNP ID
-
hCV32125434
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128954139 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCGTCATACCCAGCCTATTGTTTT[A/C]TTTACTATCTTTTTTCTGTTCCAGG
- Phenotype
-
MIM: 610746
MIM: 615587
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DOLK
PubMed Links
Gene Details
- Gene
- DOLK
- Gene Name
- dolichol kinase
There are no transcripts associated with this gene.
- Gene
- NUP188
- Gene Name
- nucleoporin 188
There are no transcripts associated with this gene.
View Full Product Details