Product Details

SNP ID
rs7219982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55398876 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTCTTTCAGCACTGGGAACTGT[A/C]CGGAATTTCCATTTATCTTCCAGCC
Phenotype
MIM: 604467
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MMD PubMed Links

Gene Details

Gene
MMD
Gene Name
monocyte to macrophage differentiation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012329.2 Intron NP_036461.2
XM_006721795.2 Intron XP_006721858.1

View Full Product Details