Product Details

SNP ID
rs11680521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95412296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTATACAGAAAGTTCTAACATA[A/G]AGCACTGAGTCAATGTGGGCACTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAHD2A PubMed Links

Gene Details

Gene
FAHD2A
Gene Name
fumarylacetoacetate hydrolase domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016044.2 Intron NP_057128.2
XM_011511282.2 Intron XP_011509584.1
XM_011511283.2 Intron XP_011509585.1
XM_011511284.2 Intron XP_011509586.1
XM_011511285.2 Intron XP_011509587.1
XM_011511286.2 Intron XP_011509588.1
XM_011511287.2 Intron XP_011509589.1
XM_011511288.2 Intron XP_011509590.1
XM_011511289.2 Intron XP_011509591.1
XM_011511290.2 Intron XP_011509592.1
XM_017004236.1 Intron XP_016859725.1
XM_017004237.1 Intron XP_016859726.1
XM_017004238.1 Intron XP_016859727.1
XM_017004239.1 Intron XP_016859728.1
XM_017004240.1 Intron XP_016859729.1
XM_017004241.1 Intron XP_016859730.1
XM_017004242.1 Intron XP_016859731.1
XM_017004243.1 Intron XP_016859732.1
XM_017004244.1 Intron XP_016859733.1
XM_017004245.1 Intron XP_016859734.1
XM_017004246.1 Intron XP_016859735.1
XM_017004247.1 Intron XP_016859736.1
XM_017004248.1 Intron XP_016859737.1
XM_017004249.1 Intron XP_016859738.1
XM_017004250.1 Intron XP_016859739.1

View Full Product Details