Product Details

SNP ID
rs4812806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:44257543 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCCAGAGCCCCAGAGCCACAGGC[A/C]TAGCTACTGATCCATACAGAGACCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GDAP1L1 PubMed Links

Gene Details

Gene
GDAP1L1
Gene Name
ganglioside induced differentiation associated protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256737.1 Intron NP_001243666.1
NM_001256738.1 Intron NP_001243667.1
NM_001256739.1 Intron NP_001243668.1
NM_001256740.1 Intron NP_001243669.1
NM_024034.4 Intron NP_076939.3

View Full Product Details