Product Details

SNP ID
rs8003859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:45125406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTGGAGGCAGAATCACCAGAAG[G/T]TGGTAATAACTACATATGAAGCAGT
Phenotype
MIM: 186947 MIM: 614907
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FKBP3 PubMed Links

Gene Details

Gene
FKBP3
Gene Name
FK506 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002013.3 Intron NP_002004.1
XM_011536565.2 Intron XP_011534867.1
XM_017021092.1 Intron XP_016876581.1
XM_017021093.1 Intron XP_016876582.1
Gene
PRPF39
Gene Name
pre-mRNA processing factor 39
There are no transcripts associated with this gene.

View Full Product Details