Product Details

SNP ID
rs3135545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:33819760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGAATGTATATTTTGGACATGA[A/G]TTTAATATAATGGTCTTATGTGTTA
Phenotype
MIM: 600405
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RFC3 PubMed Links

Gene Details

Gene
RFC3
Gene Name
replication factor C subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002915.3 Intron NP_002906.1
NM_181558.2 Intron NP_853536.2
XM_011535172.2 Intron XP_011533474.1
XM_011535173.2 Intron XP_011533475.1
XM_011535174.2 Intron XP_011533476.1
XM_011535175.1 Intron XP_011533477.1
XM_017020680.1 Intron XP_016876169.1
XM_017020681.1 Intron XP_016876170.1
XM_017020682.1 Intron XP_016876171.1
XM_017020683.1 Intron XP_016876172.1

View Full Product Details