Product Details

SNP ID
rs34223855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:498640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTGAACCACCCAGCACACACAT[C/G]GCTGCATAGGAGGAAAACAGGGTGA
Phenotype
MIM: 173430
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDGFA PubMed Links

Gene Details

Gene
PDGFA
Gene Name
platelet derived growth factor subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002607.5 Intron NP_002598.4
NM_033023.4 Intron NP_148983.1
XM_011515415.1 Intron XP_011513717.1
XM_011515416.1 Intron XP_011513718.1
XM_011515417.1 Intron XP_011513719.1
XM_011515418.1 Intron XP_011513720.1
XM_011515419.2 Intron XP_011513721.1
XM_017012289.1 Intron XP_016867778.1

View Full Product Details