Product Details

SNP ID
rs16852013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:128880300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCTCAGAAATCTGGCTCCAAGA[C/T]GCTTCCCAAGGGTGGCACAACCCAG
Phenotype
MIM: 611103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAD9 PubMed Links

Gene Details

Gene
ACAD9
Gene Name
acyl-CoA dehydrogenase family member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014049.4 Intron NP_054768.2
XM_017006264.1 Intron XP_016861753.1
XM_017006265.1 Intron XP_016861754.1
Gene
LOC653712
Gene Name
intraflagellar transport 122 homolog (Chlamydomonas) pseudogene
There are no transcripts associated with this gene.

View Full Product Details