Product Details

SNP ID
rs16861035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:48137748 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCTGATAGATCTTTTTACTGCC[C/T]GTGAAATCAAGTCCTGCAAATTTGT
Phenotype
MIM: 600583 MIM: 600058
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEC PubMed Links

Gene Details

Gene
TEC
Gene Name
tec protein tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003215.2 Intron NP_003206.2
XM_011513737.2 Intron XP_011512039.1
XM_011513741.1 Intron XP_011512043.1
XM_017008569.1 Intron XP_016864058.1
Gene
TXK
Gene Name
TXK tyrosine kinase
There are no transcripts associated with this gene.

View Full Product Details