Product Details

SNP ID
rs16843947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101519415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATCCCATTAGATTGTTTCACC[A/G]CGGCCTCGATGTAAAACATTTTTGG
Phenotype
MIM: 612846
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM172BP PubMed Links

Gene Details

Gene
FAM172BP
Gene Name
family with sequence similarity 172, member B pseudogene
There are no transcripts associated with this gene.

Gene
SENP7
Gene Name
SUMO1/sentrin specific peptidase 7
There are no transcripts associated with this gene.

View Full Product Details