Product Details

SNP ID
rs4996522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131425777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAGACAACACATTTCTATAAA[A/G]GAGGCCACAAACCAACTAAATCCTA
Phenotype
MIM: 610499
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAPGEF6 PubMed Links

Gene Details

Gene
RAPGEF6
Gene Name
Rap guanine nucleotide exchange factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164386.1 6501 UTR 3 NP_001157858.1
NM_001164387.1 6501 UTR 3 NP_001157859.1
NM_001164388.1 6501 UTR 3 NP_001157860.1
NM_001164389.1 6501 Intron NP_001157861.1
NM_001164390.1 6501 Intron NP_001157862.1
NM_016340.5 6501 UTR 3 NP_057424.3

View Full Product Details