Product Details

SNP ID
rs17866311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127343808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATTTAACAGGATACATCTCATG[C/T]AGAAATAATCCAGTATTGAGAAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF800 PubMed Links

Gene Details

Gene
ZNF800
Gene Name
zinc finger protein 800
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176814.4 7032 Intron NP_789784.2
XM_005250181.3 7032 Intron XP_005250238.1
XM_006715867.3 7032 UTR 3 XP_006715930.1
XM_006715868.3 7032 UTR 3 XP_006715931.1
XM_006715869.3 7032 UTR 3 XP_006715932.1
XM_011515859.2 7032 UTR 3 XP_011514161.1

View Full Product Details