Product Details

SNP ID
rs17865903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127344733 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAGTGTGTGTAGGAATTTAGTAT[G/T]TGATAAATGTGGAATTTTTAAAACA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF800 PubMed Links

Gene Details

Gene
ZNF800
Gene Name
zinc finger protein 800
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176814.4 6107 Intron NP_789784.2
XM_005250181.3 6107 Intron XP_005250238.1
XM_006715867.3 6107 UTR 3 XP_006715930.1
XM_006715868.3 6107 UTR 3 XP_006715931.1
XM_006715869.3 6107 UTR 3 XP_006715932.1
XM_011515859.2 6107 UTR 3 XP_011514161.1

View Full Product Details