Product Details

SNP ID
rs17862359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:127348236 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTCATAGGAAAACAGATCCAATT[C/T]AGGTCTCATGCAACAACTTGGTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF800 PubMed Links

Gene Details

Gene
ZNF800
Gene Name
zinc finger protein 800
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176814.4 Intron NP_789784.2
XM_005250181.3 Intron XP_005250238.1
XM_006715867.3 Intron XP_006715930.1
XM_006715868.3 Intron XP_006715931.1
XM_006715869.3 Intron XP_006715932.1
XM_011515859.2 Intron XP_011514161.1

View Full Product Details