Product Details

SNP ID
rs17857472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130173878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAAGTAAAAGTTTTTCCGTCG[C/G]GATACTTCGGATGTGGAGGTAATCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TMEM209 PubMed Links

Gene Details

Gene
TMEM209
Gene Name
transmembrane protein 209
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301163.1 1819 Missense Mutation CCC,CGC P427R NP_001288092.1
NM_032842.3 1819 Missense Mutation CCC,CGC P469R NP_116231.2
XM_005250655.1 1819 Missense Mutation CCC,CGC P468R XP_005250712.1
XM_011516646.2 1819 Missense Mutation CCC,CGC P468R XP_011514948.1

View Full Product Details