Product Details

SNP ID
rs7878827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:9469126 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAGCCACTTTACTCAGTCGACCGA[C/G]ATAGAATTATTATAATGCTTGGAGC
Phenotype
MIM: 300196
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TBL1X PubMed Links

Gene Details

Gene
TBL1X
Gene Name
transducin (beta)-like 1X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139466.1 Intron NP_001132938.1
NM_001139467.1 Intron NP_001132939.1
NM_001139468.1 Intron NP_001132940.1
NM_005647.3 Intron NP_005638.1
XM_011545571.2 Intron XP_011543873.1

View Full Product Details