Product Details

SNP ID
rs17804294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:67243758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTAACACACACATGCACAAAG[C/G]ATTCCATCATCAAACAAGGTTAGGA
Phenotype
MIM: 603922
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SUCLG2 PubMed Links

Gene Details

Gene
SUCLG2
Gene Name
succinate-CoA ligase GDP-forming beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177599.1 Intron NP_001171070.1
NM_003848.3 Intron NP_003839.2
XM_017007415.1 Intron XP_016862904.1
XM_017007416.1 Intron XP_016862905.1
XM_017007417.1 Intron XP_016862906.1
XM_017007418.1 Intron XP_016862907.1
XM_017007419.1 Intron XP_016862908.1
XM_017007420.1 Intron XP_016862909.1

View Full Product Details