Product Details
- SNP ID
-
hCV33698432
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:66430977 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAACATTAAGTTAATAGCAGAAA[A/C]CATTCCCTCAAATATTTGGCAAATA
- Phenotype
-
MIM: 612051
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BEAN1
PubMed Links
Gene Details
- Gene
- BEAN1
- Gene Name
- brain expressed, associated with NEDD4, 1
There are no transcripts associated with this gene.
View Full Product Details