Product Details
- SNP ID
-
rs16970909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:78194413 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGATTATAGGCGTGAGCCACACTTT[G/T]CCTTCTTTGCACCAGGGAGAATTAT
- Phenotype
-
MIM: 188300
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AFMID
PubMed Links
Gene Details
- Gene
- AFMID
- Gene Name
- arylformamidase
- Gene
- TK1
- Gene Name
- thymidine kinase 1
There are no transcripts associated with this gene.
View Full Product Details