Product Details

SNP ID
rs17855130
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9839167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGTGCACCTGTAGTTGACTGGA[A/G]AACACTGGGCAACAGGAAAGCCAGG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARPC4-TTLL3 PubMed Links
Additional Information
For this assay, SNP(s) [rs141208638] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
There are no transcripts associated with this gene.

Gene
LOC107986062
Gene Name
endogenous retrovirus group K member 9 Pol protein-like
There are no transcripts associated with this gene.

Gene
RPUSD3
Gene Name
RNA pseudouridylate synthase domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142547.1 733 Silent Mutation TTC,TTT F228F NP_001136019.1
NM_173659.3 733 Silent Mutation TTC,TTT F243F NP_775930.2
XM_011533627.2 733 Intron XP_011531929.1
Gene
TTLL3
Gene Name
tubulin tyrosine ligase like 3
There are no transcripts associated with this gene.

View Full Product Details