Product Details

SNP ID
rs34234562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:170901344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAATTCTAGAATAAGCACTGTA[A/C]CCACTAAGTGCTGGGAGGCACAGTA
Phenotype
MIM: 605782
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EIF5A2 PubMed Links

Gene Details

Gene
EIF5A2
Gene Name
eukaryotic translation initiation factor 5A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020390.5 Intron NP_065123.1

View Full Product Details