Product Details

SNP ID
rs17467252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:10491990 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATACACAGTAATTCTGATCAGGT[G/T]AGTTTGGATAAGTTCCTGAAGGAGT
Phenotype
MIM: 611434
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLNK PubMed Links

Gene Details

Gene
CLNK
Gene Name
cytokine dependent hematopoietic cell linker
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052964.2 1990 Intron NP_443196.2
XM_011513775.2 1990 Intron XP_011512077.1
XM_017007684.1 1990 UTR 3 XP_016863173.1
XM_017007685.1 1990 Intron XP_016863174.1

View Full Product Details