Product Details

SNP ID
rs17064302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:1831813 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACTTTTCAGTGTGTTGACTTTCA[C/T]GTCGATGACAGGCAGCTTTTTCTCA
Phenotype
MIM: 608136
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF10 PubMed Links

Gene Details

Gene
ARHGEF10
Gene Name
Rho guanine nucleotide exchange factor 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308152.1 Intron NP_001295081.1
NM_001308153.1 Intron NP_001295082.1
NM_014629.3 Intron NP_055444.2
XM_005266041.3 Intron XP_005266098.1
XM_011534767.2 Intron XP_011533069.1
XM_011534768.1 Intron XP_011533070.1
XM_011534770.2 Intron XP_011533072.1
XM_017014003.1 Intron XP_016869492.1

View Full Product Details