Product Details
- SNP ID
-
rs17191079
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:95091354 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTTCTTTCTCTTCATCCTCCTC[A/G]GATCTCCTAAGCTATTACAGAGGGA
- Phenotype
-
MIM: 606241
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DICER1
PubMed Links
Gene Details
- Gene
- DICER1
- Gene Name
- dicer 1, ribonuclease III
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001195573.1 |
5497 |
Intron |
|
|
NP_001182502.1 |
NM_001271282.2 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
NP_001258211.1 |
NM_001291628.1 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
NP_001278557.1 |
NM_030621.4 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
NP_085124.2 |
NM_177438.2 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
NP_803187.1 |
XM_011536599.1 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
XP_011534901.1 |
XM_011536600.2 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
XP_011534902.1 |
XM_011536601.2 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
XP_011534903.1 |
XM_011536602.2 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
XP_011534904.1 |
XM_011536604.2 |
5497 |
Silent Mutation |
TCC,TCT |
S1657S |
XP_011534906.1 |
XM_011536605.2 |
5497 |
Silent Mutation |
TCC,TCT |
S1299S |
XP_011534907.1 |
XM_017021120.1 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
XP_016876609.1 |
XM_017021121.1 |
5497 |
Silent Mutation |
TCC,TCT |
S1792S |
XP_016876610.1 |
XM_017021122.1 |
5497 |
Silent Mutation |
TCC,TCT |
S1657S |
XP_016876611.1 |
XM_017021123.1 |
5497 |
Silent Mutation |
TCC,TCT |
S1657S |
XP_016876612.1 |
View Full Product Details