Product Details
- SNP ID
-
rs17340540
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:31125065 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACACCATACACCTCAGGTAGTCAAG[A/T]CTGCAAATGCCTATATAACTTCAAT
- Phenotype
-
MIM: 300377
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DMD
PubMed Links
Gene Details
- Gene
- DMD
- Gene Name
- dystrophin
View Full Product Details