Product Details
- SNP ID
-
rs4987365
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:169694248 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGCTGTCTCAAGAAAGCTTAGCA[C/G]TTTCCAACACAAAAATGTTCAATAA
- Phenotype
-
MIM: 153240
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SELL
PubMed Links
Gene Details
- Gene
- SELL
- Gene Name
- selectin L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000655.4 |
|
Intron |
|
|
NP_000646.2 |
View Full Product Details