Product Details

SNP ID
rs11554959
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32889069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGATCTACGCCAACTTCTTTCC[C/G]TATGGTGACGCCTCCAAGTTTGCCG
Phenotype
MIM: 142622
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HPCA PubMed Links

Gene Details

Gene
HPCA
Gene Name
hippocalcin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002143.2 683 Silent Mutation CCC,CCG P57P NP_002134.2
XM_005270792.2 683 Silent Mutation CCC,CCG P57P XP_005270849.1
XM_017001118.1 683 Silent Mutation CCC,CCG P57P XP_016856607.1
Gene
TMEM54
Gene Name
transmembrane protein 54
There are no transcripts associated with this gene.

View Full Product Details