Product Details

SNP ID
hCV42656415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216415320 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCTTCGGGGCAGAACATTTCT[G/T]ACATCCATTCTAGCTCAGAGAGTGT
Phenotype
MIM: 606622
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SMARCAL1 PubMed Links

Gene Details

Gene
SMARCAL1
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127207.1 760 Missense Mutation GAC,TAC D206Y NP_001120679.1
NM_014140.3 760 Missense Mutation GAC,TAC D206Y NP_054859.2

View Full Product Details