Product Details
- SNP ID
-
rs7754502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:167187672 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAGCTGTGAGCCTGTGGCTTCTGT[G/T]CGGGTGTGTAAGTGCACCCAGGTCT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TCP10L2
PubMed Links
Gene Details
- Gene
- TCP10L2
- Gene Name
- t-complex 10-like 2
View Full Product Details