Product Details

SNP ID
rs11537581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47590648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCGAAGTCGCCCCCGATGTTCA[C/T]GTACACCTTGTCGAAGGTCACCGCC
Phenotype
MIM: 614911 MIM: 603846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QTNF4 PubMed Links

Gene Details

Gene
C1QTNF4
Gene Name
C1q and tumor necrosis factor related protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031909.2 430 Missense Mutation ATG,GTG M55V NP_114115.2
XM_017017165.1 430 Missense Mutation ATG,GTG M55V XP_016872654.1
XM_017017166.1 430 Missense Mutation ATG,GTG M55V XP_016872655.1
Gene
FAM180B
Gene Name
family with sequence similarity 180 member B
There are no transcripts associated with this gene.

Gene
NDUFS3
Gene Name
NADH:ubiquinone oxidoreductase core subunit S3
There are no transcripts associated with this gene.

View Full Product Details