Product Details

SNP ID
rs7964357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121893222 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGAAGGCAGGAGCAGATACAGG[C/T]TTTTAGCCCCTGGCGCATTTCTGCA
Phenotype
MIM: 609695 MIM: 603146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPD PubMed Links

Gene Details

Gene
HPD
Gene Name
4-hydroxyphenylpyruvate dioxygenase
There are no transcripts associated with this gene.

Gene
PSMD9
Gene Name
proteasome 26S subunit, non-ATPase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261400.2 Intron NP_001248329.1
NM_002813.6 Intron NP_002804.2

View Full Product Details