Product Details

SNP ID
rs11551784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78542529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCATGGAAGCTATTGGACATGC[A/C]GGCACCTGTTTGGGAATTTTAGCAA
Phenotype
MIM: 614681 MIM: 176846
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HYKK PubMed Links

Gene Details

Gene
HYKK
Gene Name
hydroxylysine kinase
There are no transcripts associated with this gene.

Gene
PSMA4
Gene Name
proteasome subunit alpha 4
There are no transcripts associated with this gene.

View Full Product Details