Product Details

SNP ID
rs8006221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73146433 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCAACTCCTGGCCTTAAGCTCT[C/T]TATTTTTTAAGCAAATTTGCACTGT
Phenotype
MIM: 104311
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSEN1 PubMed Links

Gene Details

Gene
PSEN1
Gene Name
presenilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000021.3 Intron NP_000012.1
NM_007318.2 Intron NP_015557.2
XM_005267864.2 Intron XP_005267921.1
XM_005267866.1 Intron XP_005267923.1
XM_011536971.2 Intron XP_011535273.1
XM_011536972.2 Intron XP_011535274.1
XM_011536973.1 Intron XP_011535275.1
XM_011536974.1 Intron XP_011535276.1

View Full Product Details