Product Details

SNP ID
rs4606484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66595237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGTAATAACAGCCCAGTAAATC[C/T]TAGGGGATTATGAGAAGAAGGGCTT
Phenotype
MIM: 603864
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC87 PubMed Links

Gene Details

Gene
CCDC87
Gene Name
coiled-coil domain containing 87
There are no transcripts associated with this gene.

Gene
CCS
Gene Name
copper chaperone for superoxide dismutase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005125.1 Intron NP_005116.1

View Full Product Details