Product Details
- SNP ID
-
rs11539684
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:195569050 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCCAAGCAAAATCCACGTGAAA[A/G]AGTTGGATGATGCAGGTACAGGAAT
- Phenotype
-
MIM: 107740
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOD
PubMed Links
Gene Details
- Gene
- APOD
- Gene Name
- apolipoprotein D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001647.3 |
782 |
Silent Mutation |
CTC,CTT |
L140L |
NP_001638.1 |
View Full Product Details