Product Details

SNP ID
rs808078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46660191 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGATAAGTGCTTGTGGGTTTTGT[G/T]TTTTTTTTTTTTTTTTTTTTTACAG
Phenotype
MIM: 601501
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
VPS35 PubMed Links

Gene Details

Gene
VPS35
Gene Name
VPS35 retromer complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018206.5 2771 UTR 3 NP_060676.2
XM_005256045.3 2771 UTR 3 XP_005256102.1
XM_011523227.2 2771 UTR 3 XP_011521529.1

View Full Product Details