Product Details
- SNP ID
-
rs7548312
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:161545713 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATATCGCCAGAGCTTATTCTCAC[A/G]ATCATGTCACCAAGTAATCAGACTT
- Phenotype
-
MIM: 146740
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FCGR3A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs145421193] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FCGR3A
- Gene Name
- Fc fragment of IgG receptor IIIa
View Full Product Details