Product Details

SNP ID
rs28485070
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:152882389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGATGTGGCTTTCTAAACATCA[A/G]GGAGAGTGTTTGGAGCAGGGCTGTG
Phenotype
MIM: 601148
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMCP PubMed Links

Gene Details

Gene
SMCP
Gene Name
sperm mitochondria associated cysteine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030663.2 Intron NP_109588.2

View Full Product Details