Product Details

SNP ID
rs28378300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:142614222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTGAGGAGGAAAAAGAATACA[C/T]ATGTTCTTTCTCTCAACAGCACCAT
Phenotype
MIM: 602734
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLS1 PubMed Links

Gene Details

Gene
PLS1
Gene Name
plastin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145319.1 Intron NP_001138791.1
NM_001172312.1 Intron NP_001165783.1
NM_002670.2 Intron NP_002661.2
XM_006713660.3 Intron XP_006713723.1
XM_011512900.2 Intron XP_011511202.1
XM_011512901.1 Intron XP_011511203.1
XM_011512903.2 Intron XP_011511205.1
XM_017006626.1 Intron XP_016862115.1
XM_017006627.1 Intron XP_016862116.1

View Full Product Details