Product Details
- SNP ID
-
rs28452706
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:15345676 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATTAATATTTAAATAAGGTGTTC[C/T]GTCACTCAGCAAACAGGTGTTCAAT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C1QTNF7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76819934] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C1QTNF7
- Gene Name
- C1q and tumor necrosis factor related protein 7
- Gene
- LOC101929095
- Gene Name
- uncharacterized LOC101929095
There are no transcripts associated with this gene.
View Full Product Details