Product Details
- SNP ID
-
rs28528780
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:50441293 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTAGGACGCTGAAGAAAAGGCGG[A/C]ATGGGAGGGGCCCGGCCGCCTTGCC
- Phenotype
-
MIM: 615669
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EMB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115322616] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EMB
- Gene Name
- embigin
View Full Product Details