Product Details

SNP ID
rs28465543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104763846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCAAAGCACTGCACATGCACA[C/G]GCTGAACACCTCCCCTTCCTTCTCC
Phenotype
MIM: 608871 MIM: 608872
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NIPSNAP3A PubMed Links

Gene Details

Gene
NIPSNAP3A
Gene Name
nipsnap homolog 3A
There are no transcripts associated with this gene.

Gene
NIPSNAP3B
Gene Name
nipsnap homolog 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018376.3 106 UTR 5 NP_060846.2
XM_011518839.1 106 UTR 5 XP_011517141.1
XM_017014889.1 106 UTR 5 XP_016870378.1

View Full Product Details