Product Details
- SNP ID
-
rs4135205
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:110248048 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAGTCTTTTTAAAAAAAGCTTA[C/T]AGACTAAGGATATAAAGAAAATATT
- Phenotype
-
MIM: 187700
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TXN
PubMed Links
Gene Details
- Gene
- TXN
- Gene Name
- thioredoxin
View Full Product Details