Product Details

SNP ID
rs28625019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:123423016 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCCTTGAGTTATTATTTGGCGG[A/G]ACCGGGGGGCAGCAAGGTGACACTT
Phenotype
MIM: 614093
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RILPL2 PubMed Links

Gene Details

Gene
RILPL2
Gene Name
Rab interacting lysosomal protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145058.2 Intron NP_659495.1
XM_011538012.2 Intron XP_011536314.1
XM_017018959.1 Intron XP_016874448.1

View Full Product Details