Product Details
- SNP ID
-
rs28422946
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31441235 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGAAGGTGCCCGGCCCAGTGCTTC[A/G]GGCCACCCTAGGCCCTACCTACCTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF843
PubMed Links
Gene Details
- Gene
- ZNF843
- Gene Name
- zinc finger protein 843
View Full Product Details