Product Details

SNP ID
rs28418559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42839898 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGAGGATCTCTTTAACTCCAAT[G/T]GGTGACTGGTTGGAGGATTCCCCAT
Phenotype
MIM: 176399
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PSG10P PubMed Links

Gene Details

Gene
PSG10P
Gene Name
pregnancy specific beta-1-glycoprotein 10, pseudogene
There are no transcripts associated with this gene.

View Full Product Details