Product Details
- SNP ID
-
rs28429169
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57532343 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAAAGGATTTATTATATTCATAG[A/G]TCCTCATAGAGGGTGGCACAGCAAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF549
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116183275] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF549
- Gene Name
- zinc finger protein 549
- Gene
- ZNF550
- Gene Name
- zinc finger protein 550
There are no transcripts associated with this gene.
View Full Product Details