Product Details

SNP ID
rs28915402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:572293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTTTCAACGCTTCAACCCCCT[C/G]GGGCGCACCGCTCTGCGCTCATTGC
Phenotype
MIM: 109480
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BSG PubMed Links

Gene Details

Gene
BSG
Gene Name
basigin (Ok blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322243.1 Intron NP_001309172.1
NM_001728.3 Intron NP_001719.2
NM_198589.2 Intron NP_940991.1
NM_198590.2 Intron NP_940992.1
NM_198591.2 Intron NP_940993.1
XM_017027173.1 Intron XP_016882662.1

View Full Product Details