Product Details

SNP ID
rs28400393
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:206903825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCACCTGGATGTACTATCCAATC[G/T]GTGATGACATTCCCTGCTAATAAAA
Phenotype
MIM: 606015 MIM: 604136
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142473.1 1307 UTR 3 NP_001135945.1
NM_001193338.1 1307 UTR 3 NP_001180267.1
NM_005449.4 1307 UTR 3 NP_005440.1
XM_005273351.3 1307 UTR 3 XP_005273408.1
XM_005273352.4 1307 Intron XP_005273409.1
Gene
IL24
Gene Name
interleukin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185156.1 1307 UTR 3 NP_001172085.1
NM_001185157.1 1307 UTR 3 NP_001172086.1
NM_001185158.1 1307 UTR 3 NP_001172087.1
NM_006850.3 1307 UTR 3 NP_006841.1
XM_011509101.2 1307 UTR 3 XP_011507403.1
XM_017000121.1 1307 UTR 3 XP_016855610.1

View Full Product Details