Product Details

SNP ID
rs28470757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:65091162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAGGAGGGAGGGGAAATAGTTTA[C/G]TTTTCATAACTGTGGAGATTATCAG
Phenotype
MIM: 616889 MIM: 179508
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP68 PubMed Links

Gene Details

Gene
CEP68
Gene Name
centrosomal protein 68
There are no transcripts associated with this gene.

Gene
RAB1A
Gene Name
RAB1A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004161.4 Intron NP_004152.1
NM_015543.1 Intron NP_056358.1
XM_005264468.1 Intron XP_005264525.1

View Full Product Details